Learn More
Medchemexpress LLC Propafenone hydrochloride | 34183-22-7 | 98.5% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0432A10MM/1ML
Encompass_Preferred
Propafenone hydrochloride is an anti-arrhythmic medication primarily used to treat rapid heartbeats, including atrial and ventricular arrhythmias. It functions by slowing the influx of sodium ions into cardiac muscle cells, thereby reducing their excitability. This compound is notably more selective for cells exhibiting a high heart rate and also possesses beta-adrenergic blocking activity.
  • Treats rapid heartbeats like atrial and ventricular arrhythmias.
  • Slows sodium ion influx into cardiac muscle cells.
  • Decreases cellular excitability.
  • Selective for cells with high heart rates.
  • Exhibits beta-adrenergic blocking activity.
Catalog No. 50-004-07895