Learn More
Medchemexpress LLC Propanoic acid, 2-(diethoxyphosphinyl)-, ethyl ester | 3699-66-9 | 99.4% | 238.22 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW013436100G
Encompass_Preferred
Ethyl 2-(diethoxyphosphoryl)propanoate is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is a Wittig-Horner reagent that condenses with a wide variety of aldehydes and ketones to form acrylates.
  • Can be used as a biological material or organic compound
  • Suitable for life science related research
  • Condenses with a wide variety of aldehydes and ketones
  • Forms acrylates
Catalog No. 50-004-54618