Learn More
Medchemexpress LLC Propanoic acid, 2-hydroxy-, phenylmethyl ester | 2051-96-9 | MFCD00157065 | 95.0% | 180.20 g/mol | C10H12O3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW016484100MG
Encompass_Preferred
Benzyl lactate (benzyl 2-hydroxypropanoate) is a lactate ester used as a small-molecule reagent and intermediate in organic synthesis, flavor and fragrance formulation, and chiral synthesis. It is a colorless to light-yellow liquid (C10H12O3, Mw 180.20 g/mol) with moderate density and solvent properties suitable for laboratory research.
  • Lactate ester useful as an intermediate in organic synthesis.
  • Suitable for flavor and fragrance applications due to a mild, clean odor.
  • Applicable in chiral synthesis and as a building-block reagent.
  • Available in small pack sizes for laboratory research.
  • Typical purity 95.0%.
  • Provided with COA, SDS, and analytical data for traceability.
Catalog No. 50-003-01797