Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propanoic acid, 3-[[13-(4-formylphenyl)-13-oxo-3,6,9-trioxa-12-azatridec-1-yl]oxy]- | 1309460-27-2 | MFCD18916985 | 99.1% | 397.42 g/mol | C19H27NO8 | 100 MG
SDP

Supplier:  Medchemexpress LLC HY130097100MG

Encompass_Preferred

Ald-Ph-amido-PEG4-C2-acid is a non-cleavable PEG4-based linker designed for use in antibody-drug conjugate (ADC) research. Supplied as a powder for conjugation experiments and linker development.

  • Non-cleavable PEG4 linker for stable antibody-payload conjugation.
  • Aldehyde functional group suitable for oxime or hydrazone conjugation chemistry.
  • Purity 99.1% (LCMS).
  • Molecular weight 397.42 g/mol and formula C19H27NO8.
  • Soluble to 10 mM in DMSO.
  • Store as a powder at -4°C under nitrogen.
  • Available as 100 MG powder for research applications.

Catalog No. 50-002-02142


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.