Learn More
Medchemexpress LLC Propanoic acid, 3-[[19-(4-formylphenyl)-19-oxo-3,6,9,12,15-pentaoxa-18-azanonadec-1-yl]oxy]- | 2055013-55-1 | 95.3% | 1 G

Supplier: Medchemexpress LLC HY1304961G
Ald-Ph-PEG6-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are composed of two distinct ligands linked by an intermediary; one ligand binds to an E3 ubiquitin ligase, and the other targets a specific protein. This mechanism allows PROTACs to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
- PEG-based PROTAC linker
- Used in the synthesis of PROTACs
- Exploits the intracellular ubiquitin-proteasome system
- Selectively degrades target proteins
- Molecular Weight: 485.52
- Molecular Formula: C23H35NO10
- Appearance: Viscous liquid
- Color: Colorless to light yellow
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.