Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propanoic acid, 3-[2-[2-[(4-formylbenzoyl)amino]ethoxy]ethoxy]-, 1,1-dimethylethyl ester | 1807521-09-0 | 97.0% | 25 MG
SDP

Supplier:  Medchemexpress LLC 25MGHY130202

Encompass_Preferred

This PEG-based PROTAC linker can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • Molecular weight: 365.42
  • Chemical formula: C19H27NO6
  • Appearance: Colorless to light yellow viscous liquid
  • Purity: 97.0% (NMR)
  • Storage at 4°C under nitrogen
  • Stable for 6 months at -80°C in solvent, or 1 month at -20°C in solvent under nitrogen
  • Soluble in DMSO (≥ 100 mg/mL)
  • Used as a PROTAC linker

Catalog No. 50-003-42727


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.