Learn More
Medchemexpress LLC Propanoic acid, 3-[2-[2-[(4-formylbenzoyl)amino]ethoxy]ethoxy]-, 2,5-dioxo-1-pyrrolidinyl ester | 1807521-07-8 | >95.0% | 406.39 g/mol | C19H22N2O8 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY130104500MG
Encompass_Preferred
Ald-Ph-amido-PEG2-C2-NHS ester is a nonclaevable 2-unit PEG linker for antibody-agent-conjugation (ADC)
Catalog No. 50-002-04285