Learn More
Medchemexpress LLC Propanoic acid, 3-[2-[2-[(4-formylbenzoyl)amino]ethoxy]ethoxy]-, 2,5-dioxo-1-pyrrolidinyl ester | 1807521-07-8 | 98.5% | 406.39 | C19H22N2O8 | 250 MG

Supplier: Medchemexpress LLC HY130104250MG
Ald-Ph-amido-PEG2-C2-NHS ester is a noncleavable two-unit polyethylene glycol (PEG) linker designed for antibody-drug conjugation (ADC). It features an activated N-hydroxysuccinimide (NHS) ester for amine coupling and a para-formylbenzoyl aldehyde for conjugation chemistry. The product is supplied as a colorless to light-yellow liquid with high purity and is intended for research use.
- Noncleavable two-unit PEG linker suitable for ADC construction.
- NHS ester for efficient amine coupling.
- Aldehyde (formyl) functional group for site-specific conjugation.
- High purity (approximately 98.5%).
- Liquid, colorless to light yellow appearance.
- Molecular weight 406.39 and formula C19H22N2O8.
- Storage: pure form -20°C; in solvent -80°C for 6 months, -20°C for 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.