Learn More
Medchemexpress LLC Propanoic acid, 3-[2-(2-azidoethoxy)ethoxy]-, 1,1-dimethylethyl ester | 1271728-79-0 | 95.0% | C11H21N3O4 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1407761G
Encompass_Preferred
Azido-PEG2-C2-Boc is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • Peg-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-55495