Learn More
Medchemexpress LLC Propanoic acid, 3-[2-(2-azidoethoxy)ethoxy]- | 1312309-63-9 | 99.88% | C7H13N3O4 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY140452
Encompass_Preferred
Azido-PEG2-C2-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is a click chemistry reagent containing an Azide group, capable of undergoing copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with Alkyne groups and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Contains an Azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-52560