Learn More
Medchemexpress LLC Propanoic acid, 3-[2-(2-carboxyethoxy)ethoxy]-, 1-(1,1-dimethylethyl) ester | 2086688-99-3 | ≥98.0% | 262.30 | 100 MG
SDP
Supplier:  Medchemexpress LLC 100MGHY140480
Encompass_Preferred
Acid-PEG2-C2-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. This product exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins and is for research use only.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • For research use only
Catalog No. 50-003-64778