Learn More
Medchemexpress LLC Propanoic acid, 3,3'-[1,2-ethanediylbis(oxy)]bis- | 19364-66-0 | 98.0% | 206.19 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY112559
Encompass_Preferred
Bis-PEG2-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins by containing two different ligands connected by a linker; one for an E3 ubiquitin ligase and the other for the target protein. It is for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
Catalog No. 50-004-39159