Learn More
Medchemexpress LLC Propanoic acid, 3-[[[3-[2-(7-chloro-2-quinolinyl)ethenyl]phenyl][[3-(dimethylamino)-3-oxopropyl]thio]methyl]thio]-, sodium salt, (E)- | 115103-85-0 | 99.8% | 537.07 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1998910MM/1ML
Encompass_Preferred
MK-571 sodium is an orally active, potent, and selective competitive leukotriene D4 (LTD4) receptor antagonist. It has Ki values of 0.22 and 2.1 nM in guinea pig and human lung membranes, respectively. It also acts as an inhibitor of multidrug resistance-associated protein MRP4 (ABCC4) and ABCC1 (MRP1), and inhibits constitutive and antigen-stimulated S1P (sphingosine-1-phosphate) release.
  • Orally active
  • Potent and selective competitive leukotriene D4 (LTD4) receptor antagonist
  • Inhibits multidrug resistance-associated protein MRP4 (ABCC4) and ABCC1 (MRP1)
  • Inhibits constitutive and antigen-stimulated S1P (sphingosine-1-phosphate) release
  • Available for research use only
Catalog No. 50-004-04868