Learn More
Medchemexpress LLC Propargyl-PEG1-acid | 55683-37-9 | 97.0% | 1 G
SDP
Supplier:  Medchemexpress LLC HY1305041G
Encompass_Preferred
Propargyl-PEG1-acid is a PEG-based PROTAC linker. It is utilized in the synthesis of BTK-CRBN PROTACs, including Ibrutinib-based PROTAC 4 and PROTAC 5. PROTAC 5 causes the degradation of BTK and induces the degradation of CSK, LYN, and LAT2 at 10 μM. This product also functions as a click chemistry reagent, containing an alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules.
  • Functions as a PEG-based PROTAC linker
  • Utilized in the synthesis of BTK-CRBN PROTACs
  • Acts as a click chemistry reagent
  • Contains an alkyne group for copper-catalyzed azide-alkyne cycloaddition
Catalog No. 50-003-40433