Learn More
Apexbio Technology LLC Propargyl-PEG2-acid 1859379-85-3 5g
SDP
Supplier:  Apexbio Technology LLC BA24485000
Encompass_Preferred
Propargyl-PEG2-acid (CAS 1859379-85-3) is a polyethylene glycol (PEG)-based small molecule featuring a terminal propargyl (alkyne) group and a carboxylic acid moiety The alkyne functionality facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAC) reactions commonly referred to as click chemistry This enables site-specific conjugation to azide-containing molecules for the synthesis of bioconjugates such as antibody-drug conjugates (ADCs) labeling probes or other functionalized biomolecules Its PEG2 spacer increases solubility and flexibility supporting its application in biomedical research requiring stable and efficient heterobifunctional linkers
Catalog No. 50-000-91359