Learn More
Medchemexpress LLC Propargyl-PEG2-MS | 943726-01-0 | 95.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY130584100MG
Encompass_Preferred
Propargyl-PEG2-MS is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups
Catalog No. 50-003-38485