Learn More
Medchemexpress LLC Propargyl-PEG24-amine | 00-00-0 | MFCD30723251 | 98.0% | C51H101NO24 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14003510MG
Encompass_Preferred
Propargyl-PEG24-amine is an alkyne-terminated polyethylene glycol (PEG24) linker bearing a primary amine. It provides a flexible, hydrophilic spacer and a terminal propargyl (alkyne) handle suitable for copper-catalyzed azide-alkyne cycloaddition (CuAAC) and other conjugation chemistries, commonly used in PROTAC synthesis and bioconjugation. Molecular formula C51H101NO24; molecular weight ~1112.34.
  • Contains a terminal propargyl (alkyne) group for efficient click chemistry.
  • Includes a primary amine for straightforward coupling and derivatization.
  • Provides a PEG24 spacer to improve solubility and reduce steric hindrance.
  • Offers a flexible, hydrophilic linker suitable for PROTAC and bioconjugation.
  • Supplied with high purity for research applications.
Catalog No. 50-002-61218