Learn More
Medchemexpress LLC Propargyl-PEG3-amine | 932741-19-0 | ≥98.0% | 187.24 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14003350MG
Encompass_Preferred
Propargyl-PEG3-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, possessing an alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups. PROTACs use the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Contains an alkyne group for CuAAc reactions with azide groups
Catalog No. 50-003-49855