Learn More
Medchemexpress LLC Propargyl-PEG3-methane | 89635-82-5 | 98.0% | 202.25 | 10 G
SDP
Supplier:  Medchemexpress LLC HY14005910G
Encompass_Preferred
Propargyl-PEG3-methane is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups.
  • PEG-based PROTAC linker.
  • Utilized in the synthesis of PROTACs.
  • Click chemistry reagent.
  • Contains an alkyne group.
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing azide groups.
  • Selectively degrades target proteins by exploiting the intracellular ubiquitin-proteasome system.
Catalog No. 50-003-50273