Learn More
Medchemexpress LLC Propargyl-PEG3-methane | 89635-82-5 | 98.0% | 202.25 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140059100MG
Encompass_Preferred
Propargyl-PEG3-methane is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Alkyne group that can engage in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an Alkyne group
  • Engages in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
  • Leverages the intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-49849