Learn More
Medchemexpress LLC Propargyl-PEG4-5-nitrophenyl carbonate | 1422540-81-5 | 98.2% | 397.38 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140048100MG
Encompass_Preferred
Propargyl-PEG4-5-nitrophenyl carbonate is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an alkyne group, capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups. This compound is instrumental in the development of PROTACs which exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Functions as a PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent with an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Supports targeted protein degradation via the ubiquitin-proteasome system
Catalog No. 50-003-50304