Learn More
eMolecules​ 1415800-32-6 | Propargyl-PEG4-acid | Broadpharm | MFCD22683284 | 260.286 | C12H20O6 | 0.000 | OC(=O)CCOCCOCCOCCOCC#C | 1g | 200183276
Supplier:  eMolecules​ 200183276
Encompass_Preferred
Propargyl-PEG4-acid | Broadpharm | 1415800-32-6 | MFCD22683284 | 260.286 | C12H20O6 | 0.000 | OC(=O)CCOCCOCCOCCOCC#C | 1g | 200183276
Catalog No. NC1540627