Learn More
Medchemexpress LLC Propargyl-PEG4-amine 250mg | 1013921-36-2 | 250 MG

Supplier: Medchemexpress LLC HY114670250MG
Propargyl-PEG4-amine is a PEG4-based linker bearing a terminal propargyl (alkyne) group and a primary amine. It is used as a click-chemistry reagent for copper-catalyzed azide-alkyne cycloaddition (CuAAC) and as a spacer in PROTAC and bioconjugation synthesis. The material is supplied as a light yellow to brown liquid with high reported purity and is soluble in DMSO.
- Suitable for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
- Contains a PEG4 spacer to improve solubility and flexibility.
- Terminal alkyne and primary amine enable dual functionalization strategies.
- High reported purity supports synthetic and analytical workflows.
- Available in multiple package sizes to support different scales of work.
- Soluble in DMSO and compatible with common stock solution preparation and storage conditions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.