Learn More
Medchemexpress LLC Propargyl-PEG4-CH2COOH | 1429934-37-1 | >=97.0% | 290.31 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140023100MG
Encompass_Preferred
Propargyl-PEG4-CH2COOH is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This compound is also a click chemistry reagent, featuring an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • PEG-based PROTAC linker for PROTAC synthesis.
  • Click chemistry reagent.
  • Contains an Alkyne group.
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Azide groups.
Catalog No. 50-003-49475