Learn More
Medchemexpress LLC Propargyl-PEG4-CH2COOH | 1429934-37-1 | >=97.0% | 290.31 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14002350MG
Encompass_Preferred
Propargyl-PEG4-CH2COOH is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Alkyne group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups. PROTACs themselves consist of two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets the protein of interest, enabling selective degradation of target proteins via the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an Alkyne group
  • Facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups
  • Enables selective degradation of target proteins
Catalog No. 50-003-50302