Learn More
Medchemexpress LLC Propargyl-PEG4-GGFG-DXd | 2762518-94-3 | 100.0% | 1083.12 g/mol | C54H63FN8O15 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15675550MG
Encompass_Preferred
Propargyl-PEG4-GGFG-DXd is a drug-linker conjugate for antibody-drug conjugate (ADC) research that combines a cleavable GGFG-PEG4 linker with a propargyl reactive group and the DNA topoisomerase I inhibitor DXd as the payload. This compound is supplied for research use only and is intended for conjugation and ADC development studies.
  • High stated purity suitable for research applications
  • Contains a propargyl functional group for click chemistry conjugation
  • Includes a cleavable GGFG linker designed for payload release
  • Payload is a topoisomerase I inhibitor (DXd) for cytotoxic activity
  • Available in multiple pack sizes for small-scale research
  • Supplied for research use only, not for human use
Catalog No. 50-002-04431