Learn More
Selleck Chemical LLC Propargyl-PEG4-NHS ester S0655-1G
SDP
Supplier:  Selleck Chemical LLC S06551G
Encompass_Preferred
Propargyl-PEG4-NHS ester is a 4-unit amine reactive PEG linker for antibody-drug-conjugation (ADC) which is noncleavable
Catalog No. 50-000-42574