Learn More
Medchemexpress LLC Propargyl-PEG4-O-C1-NHS ester | 1161883-51-7 | 100 MG

Supplier: Medchemexpress LLC HY130390100MG
Propargyl-PEG4-O-C1-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an alkyne group capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAC) with molecules containing azide groups.
- Target PROTAC linkers
- Part of the PROTAC pathway
- Functions as a click chemistry reagent
- Contains an alkyne group for copper-catalyzed azide-alkyne cycloaddition
- Used in the synthesis of PROTACs
- Utilizes the intracellular ubiquitin-proteasome system to selectively degrade target proteins
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.