Learn More
Medchemexpress LLC Propargyl-PEG5-NHS ester | 1393330-40-9 | 98.0% | 401.41 g/mol | C18H27NO9 | 50 MG

Supplier: Medchemexpress LLC HY13038850MG
Propargyl-PEG5-NHS ester is a PEG-based clickable linker (N-hydroxysuccinimide ester) containing a terminal propargyl (alkyne) group and an NHS-ester reactive handle for coupling to primary amines. It is intended for bioconjugation workflows such as ADC and PROTAC linker synthesis.
- Contains terminal propargyl group for copper-catalyzed azide-alkyne cycloaddition.
- Features an NHS-ester reactive group for efficient coupling to primary amines.
- Suitable for ADC, PROTAC, and general bioconjugation applications.
- High purity (98.0% by NMR) ensuring reliable coupling reactions.
- Molecular weight 401.41 g/mol; formula C18H27NO9.
- Store protected from light at -20°C; in solvent store at -80°C for long-term stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.