Learn More
Medchemexpress LLC Propargyl-PEG6-alcohol | 944560-99-0 | 97.0% | 5 G
SDP
Supplier:  Medchemexpress LLC HY1303825G
Encompass_Preferred
Propargyl-PEG6-alcohol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent, containing an Alkyne group, and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an alkyne group for CuAAc reactions
  • Can react with azide-containing molecules
Catalog No. 50-003-39856