Learn More
Medchemexpress LLC Propargyl-PEG7-NHS ester | 2093152-77-1 | 489.51 g/mol | C22H35NO11 | 250 MG
SDP
Supplier:  Medchemexpress LLC HY130381250MG
Encompass_Preferred
Propargyl-PEG7-NHS ester is an amine-reactive PEG-based linker used for bioconjugation, antibody-drug conjugate (ADC) and PROTAC synthesis, and click-chemistry applications. The reagent contains a terminal propargyl (alkyne) group and an N-hydroxysuccinimide (NHS) ester for coupling to primary amines.
  • Amine-reactive NHS ester for efficient coupling to primary amines.
  • Terminal alkyne enables copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • PEG7 spacer improves solubility and provides flexibility between conjugated moieties.
  • Available in small laboratory pack sizes for research use.
Catalog No. 50-002-03377