Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propargyl-PEG8-NH2 | 1196732-52-1 | MFCD28334507 | >98.0% | 407.50 g/mol | C19H37NO8 | 50 MG
SDP

Supplier:  Medchemexpress LLC HY13018250MG

Encompass_Preferred

Propargyl-PEG8-NH2 is a heterobifunctional PEG8 linker featuring a terminal propargyl (alkyne) group and a primary amine; it is used for click chemistry (CuAAC) and amine derivatization in bioconjugation workflows such as PROTAC and ADC synthesis.

  • Contains terminal propargyl (alkyne) group for copper-catalyzed azide-alkyne cycloaddition.
  • Contains a primary amine for further derivatization and amide or carbamate coupling.
  • Formula C19H37NO8 and molecular weight 407.50 g/mol.
  • High purity, specified as ≥98.0%.
  • Common applications include ADC linkers, PROTAC linkers, and general bioconjugation building blocks.
  • CAS number 1196732-52-1 for chemical identification.

Catalog No. 50-002-02254


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.