Learn More
Broadpharm Propargyl-PEG8-NHS ester | 2182601-74-5 | MFCD31536764 | 500mg
Supplier:  Broadpharm BP23778500MG
Encompass
Propargyl-PEG8-NHS ester | Putity: 98% | MW: 533.6 | 2182601-74-5 | MFCD31536764 | 500mg
Catalog No. NC1676386