Learn More
Medchemexpress LLC Propargyl-PEG9-amine | 2093153-98-9 | 97.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY130373100MG
Encompass_Preferred
Propargyl-PEG9-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Alkyne group capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that possess Azide groups.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Reacts with molecules possessing azide groups
Catalog No. 50-003-41042