Learn More
Medchemexpress LLC Propazine-d14 | 1219802-87-5 | 99.0% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB1882S5MG
Encompass_Preferred
Propazine-d14 is a deuterium labeled Propazine, intended for research use only. It can serve as a tracer or an internal standard for quantitative analysis in various analytical techniques such as NMR, GC-MS, or LC-MS. Stable heavy isotopes like deuterium are incorporated into drug molecules primarily for quantitation during drug development and can also influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Acts as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes to aid in drug development.
  • Potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-12280