Learn More
Alkali Scientific ProPette REACH™, Long-Neck Pipette Controller, with QuickStand™, 115V, 1/pk
SDP
Supplier:  Alkali Scientific P7950
Encompass
ProPette REACH™ brings a new level of user comfort to pipetting with serological pipettes. Its extended neck does all the reaching while the user’s elbow rests on the benchtop or hood surface. ProPette REACH™ eliminates the discomfort and fatigue normally associated with aspirating and dispensing under a hood. It dramatically reduces the amount of arm lift required to aspirate and dispense in bottles, flasks, and dishes. This takes pressure off of the shoulder, elbow and wrist while minimizing the risk of repetitive strain injuries. ProPette REACH™ also includes the proprietary QuickStand™, which defaults to a neutral and out-of-the-way position but can be folded back to support the pipette at an ideal angle, acting as a virtual “pause button” during research.
Catalog No. 50-233-6223