Learn More
Medchemexpress LLC Propineb (zinc propylenebis(dithiocarbamate)) | 12071-83-9 | 1 ML

Supplier: Medchemexpress LLC HY11963010MM 1ML
Propineb (zinc propylenebis(dithiocarbamate)) is a fungicidal compound used in agricultural and research settings to control a broad spectrum of fungal plant diseases. The product is supplied in both solution and solid formats for analytical and experimental use.
- Available as 10 mM solution in DMSO, 1 mL
- Available as solid: 100 mg, 200 mg, 500 mg
- Cas number 12071-83-9
- Chemical formula C5H8N2S4Zn
- Molecular weight 289.77
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.