Learn More
Cayman Chemical PropIonIc AcId-d5 10mg
SDP
Supplier:  Cayman Chemical 3915910 MG
Encompass_Preferred
An internal standard for the quantification of propionic acid by GC- or LC-MS
Catalog No. 50-291-7279