Learn More
Selleck Chemical LLC Propiverine hydrochloride S4931-25mg
SDP
Supplier:  Selleck Chemical LLC S493125MG
Encompass_Preferred
Propiverine is an anticholinergic drug used for the treatment of overactive bladder and urinary incontinence Propiverine is a muscarinic receptor antagonist possessing additional properties i e block of L-type Ca2 channels
Catalog No. 50-311-0718