Learn More
Biotage PROPLUS LX PHYTIP COLUMNS (1ML
SDP
Supplier:  Biotage PTT918007
Encompass
ProPlus LX PhyTip columns (1mL volume) containing 80 uL affinity media per column 96/PACK
Catalog No. NC2266636