Learn More
Medchemexpress LLC Propofol-d17 β-D-glucuronide | 1683581-05-6 | 98.0% | 371.50 | C18H9D17O7 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY118774S5MG
Encompass_Preferred
Deuterium-labeled analog of propofol β-D-glucuronide supplied as a stable-isotope internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Supplied for research use only.
  • High isotopic enrichment for accurate quantitation.
  • Suitable as an internal standard for LC-MS, GC-MS, and NMR.
  • Available in small milligram quantities for trace-level studies.
  • High purity (98.0%) to minimize interfering impurities.
  • Soluble in ethanol, DMF, and DMSO for analytical preparations.
Catalog No. 50-002-26627