Learn More
Medchemexpress LLC Propranolol-d7 (ring-d7) | 344298-99-3 | MFCD01317368 | 99.8% | 266.39 g·mol⁻¹ | C16H14D7NO2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB0573S110MG
Encompass_Preferred
Propranolol-d7 (ring-d7) is the deuterium-labeled analogue of propranolol hydrochloride used as a stable-isotope internal standard for quantitative analytical methods and pharmacokinetic studies. The compound contains seven deuterium atoms on the aromatic ring, enabling accurate LC-MS/MS quantification and metabolic tracing.
  • Stable-isotope internal standard for LC-MS quantification.
  • High isotopic and chemical purity for reliable analytical performance.
  • Off-white to light yellow solid suitable for laboratory use.
  • Supplied in small vial sizes for reference and assay applications.
Catalog No. 50-002-97292