Learn More
Cayman Chemical -ProprnololhydrochlorId 25g
SDP
Supplier:  Cayman Chemical 2334925 G
Encompass_Preferred
A β-adrenergic receptor antagonist (Kds = 6.91, 0.832, and 117.49 nM for human β1-, β2-, and β3-ARs, respectively); reduces mean blood pressure, resting heart rate, and contractility and increases AV conduction time in dogs; restores normal sinus rhythm in dogs with ouabain-induced arrhythmias (ED50 = 3.8 mg/kg); acts as a non-specific serotonin receptor antagonist, reducing tranylcypromine/L-tryptophan-induced hyperactivity in rats
Catalog No. 50-299-3696