Learn More
Medchemexpress LLC Propyl gallate | 121-79-9 | 212.20 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN052410MM/1ML
Encompass_Preferred
Propyl gallate is a common food antioxidant that can inhibit the production of acrolein, glyoxal, and methylglyoxal. It also exhibits anti-inflammatory, antitumor, and cardioprotective activities. This product is for research use only and not sold to patients.
  • Common food antioxidant
  • Inhibits the production of acrolein, glyoxal, and methylglyoxal
  • Exhibits anti-inflammatory, antitumor, and cardioprotective activities
  • Increases ROS level and decreases glutathione (GSH) in vitro
  • Inhibits cancer cell growth (e.g., Calu-6 and A549), arrests the cell cycle at G1 phase, and induces apoptosis via mitochondrial and cell death receptor pathways
  • Inhibits cardiac enlargement in dilated cardiomyopathy (DCM) mice
  • Prevents left ventricular dilatation and systolic dysfunction
  • Increases myocardial contractility by enhancing myofilament sensitivity to Ca2+ in myocardial fibers
Catalog No. 50-003-90013