Learn More
Medchemexpress LLC Propyl gallate | 121-79-9 | 212.20 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN0524R
Encompass_Preferred
Propyl gallate (Standard) is the analytical standard of Propyl gallate, intended for research and analytical applications. It is a common food antioxidant and can inhibit the production of acrolein, glyoxal, and methylglyoxal.
  • Analytical standard for Propyl gallate.
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Common food antioxidant.
  • Inhibits the production of acrolein, glyoxal, and methylglyoxal.
  • Exhibits anti-inflammatory, antitumor, and cardioprotective activities.
Catalog No. 50-003-90336