Learn More
U.S. Pharmacopeia Propyl Gallate, 121-79-9, MFCD00002196, 200mg
Supplier:  U.S. Pharmacopeia 1576800
Encompass
Molecular formula C10-H12-O5, Molecular weight 212.21 g/mol, Melting Point 294.8 - 302 °F (146 - 150 °C), Synonyms: n-Propyl gallate
Catalog No. NC9664986