Learn More
Cayman Chemical PropylhexedrInhydrochlorId 5mg
SDP
Supplier:  Cayman Chemical 120585 MG
Encompass_Preferred
An α-adrenergic sympathomimetic amine that acts as a local vasoconstrictor; intended for research and forensic applications
Catalog No. 50-295-4806