Learn More
Medchemexpress LLC Propylparaben (Standard) | 94-13-3 | 180.20 | 250 MG

Supplier: Medchemexpress LLC 250MGHYN2026R
Propylparaben (Propyl parahydroxybenzoate) is an antibacterial preservative produced by plants and bacteria. It is commonly used in cosmetics, pharmaceuticals, and foods, disrupting follicular growth and steroidogenic function by altering cell cycle, apoptosis, and steroidogenic pathways. It has also been observed to reduce sperm count and motility in rats. This product is for research use only.
- Antibacterial preservative.
- High purity: 99.91%.
- Soluble in DMSO.
- Powder storage: -20°C for 3 years, 4°C for 2 years.
- In solvent storage: -80°C for 2 years, -20°C for 1 year.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.