Learn More
Cayman Chemical Propylpyrazole Triol
SDP
Supplier:  Cayman Chemical 100088411 MG
Encompass_Preferred
An ERα selective agonist with a 410-fold relative binding affinity for ERα versus ERβ
Catalog No. 50-288-8720