Learn More
Cayman Chemical PropylthIouracIl 25g
SDP
Supplier:  Cayman Chemical 1406925 G
Encompass_Preferred
An antithyroid agent; inhibits thyroid peroxidase activity in rat and monkey thyroid microsomes (IC50s = 0.081 and 4.1 μM, respectively); increases thyroid weight and serum thyroid stimulating hormone levels and decreases serum 3,5,3’-triiodothyronine and thyroxine levels in rats at 30 mg/kg; sensitivity to the bitter taste of PTU is genetically mediated and is associated with increased sensitivity to other sweet and bitter compounds
Catalog No. 50-295-5588